Assay Details
| Gene Symbol: Mtag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015480.1 | 1 - 2 | 120 | 76 | ||
| GenBank mRNA | AK042966.1 | - | 1 - 2 | 120 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lin-7 homolog B (C. elegans) | Lin7b |
LIN-7B, MALS-2, Veli2 |
Anti-sense | Chr. 7 - 45367891 - 45370564 on NCBI Build GRCm38 | |
| Downstream | small nuclear ribonucleoprotein 70 (U1) | Snrnp70 |
2700022N21Rik, 3200002N22Rik, AI325098, R74807, Rnu1p70, Rnulp70, Snrp70, Srnp70, U1-70, U1-70K |
8.5kb | Anti-sense | Chr. 7 - 45376454 - 45395742 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Mtag2
Genomic Map