Assay Details
| Gene Symbol: Vwa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035229.1 | - | 493 | 73 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Vwa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035229.1 | - | 493 | 73 | ||