Assay Details
| Gene Symbol: Gm15559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044789.1 | - | 1 - 2 | 228 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15559| Gene Symbol: Gm15559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044789.1 | - | 1 - 2 | 228 | 100 | |
Set Membership: |
|