Assay Details
| Gene Symbol: Rab3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003950.2 | NP_001003950.1 | 10 | 1577 | 71 | |
| GenBank mRNA | AK045783.1 | - | 1 | 233 | 71 | |
| AK152122.1 | - | 11 | 1724 | 71 | ||
| BC080289.1 | - | 10 | 1577 | 6816077 | 71 | |
| S79539.1 | - | 8 | 1654 | 71 | ||
More Information
Set Membership: |
|

Rab3ip
Genomic Map