Assay Details
| Gene Symbol: Hhip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020259.4 | NP_064655.4 | 13 | 4426 | 90 | |
| GenBank mRNA | AK031539.1 | - | 13 | 4423 | 90 | |
| AK083190.1 | - | 1 | 371 | 90 | ||
More Information
Set Membership: |
|

Hhip
Genomic Map