Assay Details
| Gene Symbol: Tmem41b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153525.5 | NP_705745.3 | 1 - 2 | 271 | 102 | |
| GenBank mRNA | AK029327.1 | - | 1 - 2 | 272 | 102 | |
| AK033997.1 | - | 1 - 2 | 301 | 102 | ||
| AK035266.1 | - | 2 - 3 | 116 | 102 | ||
| AK038953.1 | - | 1 - 2 | 231 | 102 | ||
| AK053588.1 | - | 1 - 2 | 272 | 102 | ||
| AK075726.1 | - | 1 - 2 | 271 | 102 | ||
| AK078357.1 | - | 1 - 2 | 272 | 102 | ||
| AK090100.1 | - | 1 - 2 | 273 | 102 | ||
| AK139961.1 | - | 1 - 2 | 272 | 102 | ||
| AK150353.1 | - | 1 - 2 | 327 | 102 | ||
| AK153048.1 | - | 1 - 2 | 327 | 102 | ||
| AK162557.1 | - | 1 - 2 | 301 | 102 | ||
| AK165226.1 | - | 1 - 2 | 272 | 102 | ||
| AK166427.1 | - | 1 - 2 | 272 | 102 | ||
| AK166455.1 | - | 1 - 2 | 272 | 102 | ||
| AK167021.1 | - | 1 - 2 | 172 | 102 | ||
| AK172876.1 | - | 1 - 2 | 290 | 102 | ||
| BC026515.1 | - | 1 - 2 | 295 | 4910818 | 102 | |
| BC027103.1 | - | 1 - 2 | 272 | 5360942 | 102 | |
| BC086759.1 | - | 724 | 102 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK035266.1 transcripts.
Set Membership: |
|

Tmem41b
Genomic Map