Assay Details
| Gene Symbol: Socs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168655.1 | NP_001162126.1 | 3 | 751 | 87 | |
| NM_001168656.1 | NP_001162127.1 | 3 | 709 | 87 | ||
| NM_001168657.1 | NP_001162128.1 | 2 | 630 | 87 | ||
| NM_007706.4 | NP_031732.1 | 3 | 778 | 87 | ||
| GenBank mRNA | AK033206.1 | - | 1 | 178 | 87 | |
| AK137403.1 | - | 3 | 658 | 87 | ||
| AK144810.1 | - | 3 | 720 | 87 | ||
| AK160537.1 | - | 3 | 735 | 87 | ||
| AK162591.1 | - | 3 | 743 | 87 | ||
| BC106153.1 | - | 3 | 749 | 4989433 | 87 | |
| U88327.1 | - | 2 | 538 | 87 | ||
More Information
Set Membership: |
|

Socs2
Genomic Map