Assay Details
| Gene Symbol: Naca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113199.1 | NP_001106670.1 | 5 - 6 | 6484 | 61 | |
| NM_001282976.1 | NP_001269905.1 | 4 - 5 | 679 | 61 | ||
| NM_013608.3 | NP_038636.2 | 4 - 5 | 568 | 61 | ||
| GenBank mRNA | AK146149.1 | - | 4 - 5 | 272 | 61 | |
| AK146197.1 | - | 4 - 5 | 306 | 61 | ||
| AK151928.1 | - | 4 - 5 | 270 | 61 | ||
| AK161678.1 | - | 4 - 5 | 263 | 61 | ||
| AK168023.1 | - | 4 - 5 | 281 | 61 | ||
| AK168169.1 | - | 4 - 5 | 270 | 61 | ||
| AK168607.1 | - | 4 - 5 | 250 | 61 | ||
| AK169432.1 | - | 4 - 5 | 340 | 61 | ||
| AK169435.1 | - | 4 - 5 | 270 | 61 | ||
| AL355707.1 | - | 4 - 5 | 301 | 61 | ||
| BC029830.1 | - | 4 - 5 | 258 | 61 | ||
| BC083340.1 | - | 4 - 5 | 253 | 5700166 | 61 | |
| BC099375.1 | - | 3 - 4 | 243 | 30525972 | 61 | |
| CX236470.1 | - | 4 - 5 | 404 | 61 | ||
| U22151.1 | - | 3 - 4 | 244 | 61 | ||
| U48364.1 | - | 5 - 6 | 6195 | 61 | ||
More Information
Set Membership: |
|

Naca
Genomic Map