Assay Details
| Gene Symbol: Pcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011045.2 | NP_035175.1 | 1 | 227 | 72 | |
| GenBank mRNA | AK012614.2 | - | 1 | 229 | 72 | |
| AK012730.1 | - | 1 | 228 | 72 | ||
| AK012932.1 | - | 1 | 227 | 72 | ||
| AK088256.1 | - | 1 | 228 | 72 | ||
| AK151238.1 | - | 1 | 228 | 72 | ||
| AK151411.1 | - | 1 | 260 | 72 | ||
| AK165217.1 | - | 1 | 228 | 72 | ||
| AK168233.1 | - | 1 | 228 | 72 | ||
| AK168596.1 | - | 1 | 231 | 72 | ||
| BC005778.1 | - | 1 | 212 | 3602797 | 72 | |
| BC010343.1 | - | 1 | 211 | 3602977 | 72 | |
| X53068.1 | - | 1 | 226 | 72 | ||
More Information
Set Membership: |
|

Pcna
Genomic Map