Assay Details
| Gene Symbol: 5830416P10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028427.1 | 7 - 8 | 2716 | 100 | ||
| GenBank mRNA | AK158034.1 | - | 7 - 8 | 2706 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

5830416P10Rik| Gene Symbol: 5830416P10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028427.1 | 7 - 8 | 2716 | 100 | ||
| GenBank mRNA | AK158034.1 | - | 7 - 8 | 2706 | 100 | |
Set Membership: |
|