Assay Details
| Gene Symbol: Sh3tc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172628.2 | NP_766216.2 | 17 | 4345 | 70 | |
| GenBank mRNA | AK052534.1 | - | 17 | 4297 | 70 | |
| AK173317.1 | - | 9 | 3278 | 70 | ||
| BC020319.2 | - | 1 | 466 | 70 | ||
| BC037000.1 | - | 5 | 1187 | 70 | ||
| BC049856.1 | - | 4 | 1025 | 70 | ||
| BC076633.1 | - | 10 | 3420 | 70 | ||
More Information
Set Membership: |
|

Sh3tc2
Genomic Map