Assay Details
| Gene Symbol: Cobll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027225.1 | NP_081501.1 | 11 | 2273 | 93 | |
| NM_177025.5 | NP_795999.2 | 12 | 2384 | 93 | ||
| GenBank mRNA | AK037010.1 | - | 13 | 2378 | 93 | |
| AK052533.1 | - | 11 | 2222 | 93 | ||
| AK135859.1 | - | 3 | 612 | 93 | ||
| AK144943.1 | - | 13 | 2431 | 93 | ||
| AK173076.1 | - | 11 | 2104 | 93 | ||
| AY308746.1 | - | 12 | 2384 | 93 | ||
| BC067007.1 | - | 11 | 2270 | 5702930 | 93 | |
More Information
Set Membership: |
|

Cobll1
Genomic Map