Assay Details
| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002504.1 | - | 1 | 72 | 95 | |
| AK078459.1 | - | 1 | 43 | 95 | ||
| AK166899.1 | - | 1 | 48 | 95 | ||
| BC046802.1 | - | 1 | 99 | 6404607 | 95 | |
| X61506.1 | - | 1 | 76 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK078459.1, AK166899.1, BC046802.1, X61506.1 transcripts.
Set Membership: |
|

Atxn10
Genomic Map