Assay Details
| Gene Symbol: Etv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023794.2 | NP_076283.2 | 13 | 3522 | 101 | |
| GenBank mRNA | AK003461.1 | - | 1 | 1016 | 101 | |
| AK049234.1 | - | 13 | 3572 | 101 | ||
| AK080674.1 | - | 13 | 3499 | 101 | ||
| AK164091.1 | - | 13 | 3572 | 101 | ||
| AK168881.1 | - | 13 | 3569 | 101 | ||
| BC034680.1 | - | 13 | 3522 | 4036564 | 101 | |
| BC039044.1 | - | 1 | 334 | 101 | ||
More Information
Set Membership: |
|

Etv5
Genomic Map