Assay Details
| Gene Symbol: Kpnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008379.3 | NP_032405.3 | 22 | 4095 | 95 | |
| GenBank mRNA | D67015.1 | - | 22 | 3823 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kpnb1| Gene Symbol: Kpnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008379.3 | NP_032405.3 | 22 | 4095 | 95 | |
| GenBank mRNA | D67015.1 | - | 22 | 3823 | 95 | |
Set Membership: |
|