Assay Details
| Gene Symbol: Chmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085498.2 | NP_001078967.1 | 8 | 1148 | 114 | |
| GenBank mRNA | AK162346.1 | - | 4 | 726 | 114 | |
| AK172123.1 | - | 6 | 1866 | 114 | ||
| BC027085.1 | - | 1 | 1263 | 114 | ||
More Information
Set Membership: |
|

Chmp6
Genomic Map