Assay Details
| Gene Symbol: Dyrk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113389.1 | NP_001106860.1 | 12 | 5128 | 89 | |
| NM_007890.2 | NP_031916.1 | 12 | 5387 | 89 | ||
| GenBank mRNA | AK009788.1 | - | 1 | 1289 | 89 | |
| AK163136.1 | - | 1 | 2223 | 89 | ||
More Information
Set Membership: |
|

Dyrk1a
Genomic Map