Assay Details
| Gene Symbol: Nisch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022656.2 | NP_073147.2 | 21 | 5283 | 89 | |
| GenBank mRNA | AF144133.1 | - | 2 | 964 | 89 | |
| AF315344.1 | - | 18 | 4562 | 89 | ||
| AK122412.1 | - | 19 | 4781 | 89 | ||
| AY032852.1 | - | 4 | 1418 | 89 | ||
| BC089329.1 | - | 16 | 4502 | 89 | ||
| BC108364.1 | - | 16 | 4477 | 89 | ||
| BP761152.1 | - | 1 | 89 | |||
More Information
Set Membership: |
|

Nisch
Genomic Map