Assay Details
| Gene Symbol: Tmem53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285812.1 | NP_001272741.1 | 1 - 2 | 211 | 71 | |
| GenBank mRNA | BC020121.1 | - | 1 - 2 | 120 | 3991677 | 71 |
More Information
Set Membership: |
|

Tmem53
Genomic Map