Assay Details
| Gene Symbol: Lin37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290569.1 | NP_001277498.1 | 4 - 5 | 490 | 101 | |
| NM_029377.2 | NP_083653.1 | 4 - 5 | 364 | 101 | ||
| GenBank mRNA | AK007858.1 | - | 4 - 5 | 365 | 101 | |
| AY428767.1 | - | 4 - 5 | 214 | 101 | ||
| BC013546.1 | - | 4 - 5 | 346 | 4210417 | 101 | |
| BY707863.1 | - | 4 - 5 | 364 | 101 | ||
More Information
Set Membership: |
|

Lin37
Genomic Map