Assay Details
| Gene Symbol: Mapre3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133350.1 | NP_579928.1 | 6 - 7 | 925 | 88 | |
| GenBank mRNA | AF468005.1 | - | 6 - 7 | 925 | 88 | |
| AK214734.1 | - | 1 - 2 | 157 | 88 | ||
| BC048833.1 | - | 5 - 6 | 1487 | 88 | ||
| BC057918.1 | - | 6 - 7 | 925 | 5134461 | 88 | |
| U51204.1 | - | 4 - 5 | 607 | 88 | ||
More Information
Set Membership: |
|

Mapre3
Genomic Map