Assay Details
| Gene Symbol: Fcrlb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029984.2 | NP_001025155.2 | 1 - 2 | 31 | 58 | |
| GenBank mRNA | AY513660.1 | - | 1 - 2 | 31 | 58 | |
| DQ119292.1 | - | 1 - 2 | 31 | 58 | ||
More Information
Set Membership: |
|

Fcrlb
Genomic Map