Assay Details
| Gene Symbol: AU015228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033197.2 | NP_001028369.1 | 1 | 2114 | 62 | |
| GenBank mRNA | AK139778.1 | - | 1 | 2115 | 62 | |
| AK163392.1 | - | 1 | 2116 | 62 | ||
More Information
Set Membership: |
|

AU015228
Genomic Map