Assay Details
| Gene Symbol: Ankrd61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025732.3 | NP_080008.1 | 2 - 3 | 393 | 93 | |
| GenBank mRNA | AK014929.1 | - | 2 - 3 | 394 | 93 | |
| AK017053.1 | - | 2 - 3 | 334 | 93 | ||
| BC052346.1 | - | 2 - 3 | 615 | 6703467 | 93 | |
More Information
Set Membership: |
|

Ankrd61
Genomic Map