Assay Details
| Gene Symbol: Morn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029112.1 | NP_083388.1 | 2 - 3 | 351 | 67 | |
| GenBank mRNA | AK077141.1 | - | 2 - 3 | 548 | 67 | |
| BC019987.1 | - | 2 - 3 | 351 | 4458628 | 67 | |
More Information
Set Membership: |
|

Morn3
Genomic Map