Assay Details
| Gene Symbol: Morn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015339.1 | - | 3 - 4 | 659 | 69 | |
| AK077141.1 | - | 3 - 4 | 703 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Morn3| Gene Symbol: Morn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015339.1 | - | 3 - 4 | 659 | 69 | |
| AK077141.1 | - | 3 - 4 | 703 | 69 | ||
Set Membership: |
|