Assay Details
| Gene Symbol: Gm5617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004191.3 | NP_001004191.2 | 1 | 523 | 65 | |
| GenBank mRNA | AK007723.1 | - | 1 | 428 | 65 | |
| BC011344.1 | - | 1 | 418 | 3481870 | 65 | |
| CN838422.1 | - | 1 | 427 | 65 | ||
More Information
Set Membership: |
|

Gm5617
Genomic Map