Assay Details
| Gene Symbol: Tubb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009450.2 | NP_033476.1 | 1 - 2 | 131 | 101 | |
| GenBank mRNA | AK134545.1 | - | 1 - 2 | 130 | 101 | |
| BC055441.1 | - | 1 - 2 | 121 | 4016867 | 101 | |
More Information
Set Membership: |
|

Tubb2a
Genomic Map