Assay Details
| Gene Symbol: Hspa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010479.2 | NP_034609.2 | 1 | 2721 | 72 | |
| GenBank mRNA | AK132331.1 | - | 1 | 2721 | 72 | |
| AK171820.1 | - | 2704 | 72 | |||
| BC054782.1 | - | 1 | 2689 | 6815894 | 72 | |
More Information
Set Membership: |
|

Hspa1a
Genomic Map