Assay Details
| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011192.3 | NP_035322.1 | 10 - 11 | 884 | 87 | |
| GenBank mRNA | AK140597.1 | - | 10 - 11 | 905 | 87 | |
| AK150939.1 | - | 10 - 11 | 904 | 87 | ||
| AK156079.1 | - | 10 - 11 | 854 | 87 | ||
| AK171348.1 | - | 10 - 11 | 900 | 87 | ||
| BC005524.1 | - | 2 - 3 | 137 | 87 | ||
| BC015255.1 | - | 10 - 11 | 884 | 4221225 | 87 | |
| BC139818.1 | - | 10 - 11 | 832 | 40141268 | 87 | |
| CT010198.1 | - | 10 - 11 | 688 | 87 | ||
| D87911.1 | - | 10 - 11 | 807 | 87 | ||
| U60330.1 | - | 10 - 11 | 688 | 87 | ||
More Information
Set Membership: |
|

Psme3
Genomic Map