Assay Details
| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011192.3 | NP_035322.1 | 1 - 2 | 234 | 73 | |
| GenBank mRNA | AK140597.1 | - | 1 - 2 | 255 | 73 | |
| AK150939.1 | - | 1 - 2 | 254 | 73 | ||
| AK156079.1 | - | 1 - 2 | 204 | 73 | ||
| AK171348.1 | - | 1 - 2 | 250 | 73 | ||
| BC015255.1 | - | 1 - 2 | 234 | 4221225 | 73 | |
| BC139818.1 | - | 1 - 2 | 182 | 40141268 | 73 | |
| D87911.1 | - | 1 - 2 | 157 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK140597.1, AK150939.1, AK156079.1, AK171348.1, BC015255.1, BC139818.1, D87911.1, NM_011192.3 transcripts.
Set Membership: |
|

Psme3
Genomic Map