Assay Details
| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011192.3 | NP_035322.1 | 3 - 4 | 336 | 103 | |
| GenBank mRNA | AK140597.1 | - | 3 - 4 | 357 | 103 | |
| AK150939.1 | - | 3 - 4 | 356 | 103 | ||
| AK156079.1 | - | 3 - 4 | 306 | 103 | ||
| AK171348.1 | - | 3 - 4 | 352 | 103 | ||
| AK182764.1 | - | 1 - 2 | 80 | 103 | ||
| AK183517.1 | - | 1 - 2 | 80 | 103 | ||
| AK184446.1 | - | 1 - 2 | 80 | 103 | ||
| AK196904.1 | - | 1 - 2 | 80 | 103 | ||
| AK198462.1 | - | 1 - 2 | 80 | 103 | ||
| AK210357.1 | - | 1 - 2 | 76 | 103 | ||
| BC015255.1 | - | 3 - 4 | 336 | 4221225 | 103 | |
| BC139818.1 | - | 3 - 4 | 284 | 40141268 | 103 | |
| CT010198.1 | - | 3 - 4 | 140 | 103 | ||
| D87911.1 | - | 3 - 4 | 259 | 103 | ||
| U60330.1 | - | 3 - 4 | 140 | 103 | ||
More Information
Set Membership: |
|

Psme3
Genomic Map