Assay Details
| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011192.3 | NP_035322.1 | 5 - 6 | 492 | 66 | |
| GenBank mRNA | AK140597.1 | - | 5 - 6 | 513 | 66 | |
| AK150939.1 | - | 5 - 6 | 512 | 66 | ||
| AK156079.1 | - | 5 - 6 | 462 | 66 | ||
| AK171348.1 | - | 5 - 6 | 508 | 66 | ||
| AK183517.1 | - | 3 - 4 | 236 | 66 | ||
| AK184446.1 | - | 3 - 4 | 236 | 66 | ||
| AK198462.1 | - | 3 - 4 | 236 | 66 | ||
| AK210357.1 | - | 3 - 4 | 232 | 66 | ||
| BC015255.1 | - | 5 - 6 | 492 | 4221225 | 66 | |
| BC139818.1 | - | 5 - 6 | 440 | 40141268 | 66 | |
| CT010198.1 | - | 5 - 6 | 296 | 66 | ||
| D87911.1 | - | 5 - 6 | 415 | 66 | ||
| U60330.1 | - | 5 - 6 | 296 | 66 | ||
More Information
Set Membership: |
|

Psme3
Genomic Map