Assay Details
| Gene Symbol: Stoml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023231.2 | NP_075720.1 | 3 - 4 | 289 | 103 | |
| GenBank mRNA | AF323178.1 | - | 3 - 4 | 309 | 103 | |
| AK002793.1 | - | 3 - 4 | 286 | 103 | ||
| BC003425.1 | - | 3 - 4 | 289 | 2609584 | 103 | |
| BC069941.1 | - | 3 - 4 | 288 | 5390357 | 103 | |
More Information
Set Membership: |
|

Stoml2
Genomic Map