Assay Details
| Gene Symbol: Stoml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023231.2 | NP_075720.1 | 9 - 10 | 941 | 57 | |
| GenBank mRNA | AF323178.1 | - | 9 - 10 | 961 | 57 | |
| AK002793.1 | - | 9 - 10 | 938 | 57 | ||
| BC003425.1 | - | 9 - 10 | 941 | 2609584 | 57 | |
| BC069941.1 | - | 9 - 10 | 940 | 5390357 | 57 | |
More Information
Set Membership: |
|

Stoml2
Genomic Map