Assay Details
| Gene Symbol: Gm4981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034869.2 | NP_001030041.1 | 1 | 318 | 64 | |
| GenBank mRNA | AK145460.1 | - | 1 | 319 | 64 | |
| BC147818.1 | - | 1 | 122 | 9087535 | 64 | |
| BC147822.1 | - | 1 | 122 | 9087539 | 64 | |
More Information
Set Membership: |
|

Gm4981
Genomic Map