Assay Details
| Gene Symbol: Gm15386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040027.2 | NP_001035116.1 | 1 - 2 | 113 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001040027.2 transcripts.
Set Membership: |
|

Gm15386
Genomic Map