Assay Details
| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024282.3 | NP_077244.1 | 4 - 5 | 846 | 92 | |
| GenBank mRNA | AK020014.1 | - | 4 - 5 | 731 | 92 | |
| AK029574.1 | - | 5 - 6 | 920 | 92 | ||
| AK049579.1 | - | 4 - 5 | 615 | 92 | ||
| AK146492.1 | - | 4 - 5 | 579 | 92 | ||
| AK160733.1 | - | 4 - 5 | 435 | 92 | ||
| BC002200.1 | - | 4 - 5 | 515 | 3489351 | 92 | |
| BC046816.1 | - | 4 - 5 | 553 | 5702476 | 92 | |
More Information
Set Membership: |
|

Desi2
Genomic Map