Assay Details
| Gene Symbol: Snrpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026943.1 | NP_081219.1 | 1 - 2 | 58 | 65 | |
| GenBank mRNA | AK007389.1 | - | 1 - 2 | 37 | 65 | |
| AK045968.1 | - | 1 - 2 | 33 | 65 | ||
| BC043014.1 | - | 1 - 2 | 58 | 5683364 | 65 | |
| BC116858.1 | - | 1 - 2 | 41 | 40126177 | 65 | |
| BC116860.1 | - | 1 - 2 | 41 | 40126179 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007389.1, AK045968.1, BC043014.1, BC116858.1, BC116860.1, NM_026943.1 transcripts.
Set Membership: |
|

Snrpd2
Genomic Map