Assay Details
| Gene Symbol: Fam83c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027788.2 | NP_082064.2 | 1 - 2 | 614 | 79 | |
| GenBank mRNA | BC113190.1 | - | 1 - 2 | 613 | 40062623 | 79 |
| BC113797.1 | - | 1 - 2 | 613 | 40062621 | 79 | |
More Information
Set Membership: |
|

Fam83c
Genomic Map