Assay Details
| Gene Symbol: Ttc30b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028235.1 | NP_082511.1 | 1 | 2421 | 99 | |
| GenBank mRNA | AK011097.1 | - | 1 | 2398 | 99 | |
| AK147134.1 | - | 1 | 2371 | 99 | ||
| BC026507.1 | - | 1 | 1613 | 99 | ||
More Information
Set Membership: |
|

Ttc30b
Genomic Map