Assay Details
| Gene Symbol: Spata25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029370.1 | NP_083646.1 | 1 - 2 | 133 | 87 | |
| GenBank mRNA | AK006145.1 | - | 1 - 2 | 108 | 87 | |
| BC116355.1 | - | 1 - 2 | 95 | 40099919 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006145.1, BC116355.1, NM_029370.1 transcripts.
Set Membership: |
|

Spata25
Genomic Map