Assay Details
| Gene Symbol: Spcs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029701.1 | NP_083977.1 | 2 - 3 | 310 | 140 | |
| GenBank mRNA | AK007432.1 | - | 2 - 3 | 289 | 140 | |
| AK028314.1 | - | 2 - 3 | 272 | 140 | ||
| AK048051.1 | - | 2 - 3 | 296 | 140 | ||
| BC019390.1 | - | 2 - 3 | 249 | 140 | ||
| BC054817.1 | - | 2 - 3 | 275 | 5375997 | 140 | |
More Information
Set Membership: |
|

Spcs3
Genomic Map