Assay Details
| Gene Symbol: Slc22a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133980.3 | NP_598741.2 | 2 - 3 | 534 | 69 | |
| GenBank mRNA | AF529220.1 | - | 2 - 3 | 527 | 69 | |
| BC021449.1 | - | 2 - 3 | 526 | 5044564 | 69 | |
More Information
Set Membership: |
|

Slc22a13
Genomic Map