Assay Details
| Gene Symbol: Acot4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134247.3 | NP_599008.3 | 2 - 3 | 799 | 76 | |
| GenBank mRNA | AK046630.1 | - | 2 - 3 | 799 | 76 | |
| AK050420.1 | - | 2 - 3 | 781 | 76 | ||
| BC113185.1 | - | 2 - 3 | 743 | 40061800 | 76 | |
More Information
Set Membership: |
|

Acot4
Genomic Map