Assay Details
| Gene Symbol: Fam104a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138598.5 | NP_613064.2 | 2 - 3 | 384 | 66 | |
| GenBank mRNA | AK009763.2 | - | 2 - 3 | 266 | 66 | |
| AK017659.1 | - | 2 - 3 | 293 | 66 | ||
| AK028386.1 | - | 2 - 3 | 297 | 66 | ||
| AK087540.1 | - | 2 - 3 | 295 | 66 | ||
| AK145119.1 | - | 2 - 3 | 204 | 66 | ||
| AK158680.1 | - | 2 - 3 | 382 | 66 | ||
| AK158872.1 | - | 2 - 3 | 430 | 66 | ||
| AK159228.1 | - | 2 - 3 | 283 | 66 | ||
| AK159659.1 | - | 2 - 3 | 283 | 66 | ||
| AV572367.1 | - | 2 - 3 | 381 | 66 | ||
| BC056429.1 | - | 2 - 3 | 125 | 66 | ||
| BC060965.1 | - | 2 - 3 | 160 | 6705115 | 66 | |
| BC089526.1 | - | 2 - 3 | 137 | 6744026 | 66 | |
More Information
Set Membership: |
|

Fam104a
Genomic Map