Assay Details
| Gene Symbol: E130308A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015681.2 | NP_001015681.1 | 1 - 2 | 284 | 81 | |
| NM_153158.5 | NP_694798.1 | 1 - 2 | 284 | 81 | ||
| GenBank mRNA | AK047478.1 | - | 1 - 2 | 285 | 81 | |
| AK047601.1 | - | 1 - 2 | 253 | 81 | ||
| AK081604.1 | - | 1 - 2 | 168 | 81 | ||
| AK087509.1 | - | 1 - 2 | 280 | 81 | ||
| BB647178.1 | - | 1 - 2 | 282 | 81 | ||
| BC017649.1 | - | 1 - 2 | 51 | 4924105 | 81 | |
| BC037070.1 | - | 1 - 2 | 102 | 5004205 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK081604.1, AK087509.1, BC017649.1, BC037070.1, NM_001015681.2, NM_153158.5 transcripts.
Set Membership: |
|

E130308A19Rik
Genomic Map