Assay Details
| Gene Symbol: Lrfn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153388.4 | NP_700437.2 | 1 - 2 | 706 | 66 | |
| GenBank mRNA | AK163603.1 | - | 1 - 2 | 564 | 66 | |
| BC023156.1 | - | 1 - 2 | 362 | 4951386 | 66 | |
| CA752061.1 | - | 1 - 2 | 706 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK163603.1, BC023156.1, NM_153388.4 transcripts.
Set Membership: |
|

Lrfn4
Genomic Map