Assay Details
| Gene Symbol: Exosc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081188.1 | NP_001074657.1 | 3 - 4 | 292 | 66 | |
| GenBank mRNA | AK011294.1 | - | 3 - 4 | 279 | 66 | |
| AK129057.1 | - | 3 - 4 | 244 | 66 | ||
| BC039655.1 | - | 3 - 4 | 277 | 66 | ||
| BC052656.1 | - | 3 - 4 | 274 | 30041585 | 66 | |
| BC094932.1 | - | 3 - 4 | 285 | 30008071 | 66 | |
| BC132363.1 | - | 3 - 4 | 307 | 40130640 | 66 | |
| BC145675.1 | - | 3 - 4 | 307 | 40130978 | 66 | |
More Information
Set Membership: |
|

Exosc7
Genomic Map