Assay Details
| Gene Symbol: Exosc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081188.1 | NP_001074657.1 | 4 - 5 | 455 | 91 | |
| GenBank mRNA | AK011294.1 | - | 4 - 5 | 442 | 91 | |
| AK129057.1 | - | 4 - 5 | 407 | 91 | ||
| BC039655.1 | - | 4 - 5 | 440 | 91 | ||
| BC052656.1 | - | 4 - 5 | 437 | 30041585 | 91 | |
| BC094932.1 | - | 4 - 5 | 448 | 30008071 | 91 | |
| BC132363.1 | - | 4 - 5 | 470 | 40130640 | 91 | |
| BC145675.1 | - | 4 - 5 | 470 | 40130978 | 91 | |
More Information
Set Membership: |
|

Exosc7
Genomic Map